Learn More
Raybiotech Inc HUMAN CYTOKINE ARRAY C5 (4)
Supplier:  Raybiotech Inc AAHCYT54
Encompass_Preferred
RayBio C-Series Human Cytokine Antibody Array 5 Kit. Detects 80 Human Cytokines. Suitable for all liquid sample types.
Catalog No. 50-104-3108