Learn More
Raybiotech Inc C-Series Human Cytokine Antibody Array 7 Kit.
Catalog No. 501948563
Encompass_Preferred
Detects 60 Human Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. View protein list on RayBiotech’s website – AAH-CYT-7-4
Catalog No. 50-194-8563 Supplier Raybiotech Inc Supplier No. AAHCYT74
Detects 60 Human Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. View protein list on RayBiotech’s website – AAH-CYT-7-4