Learn More
Raybiotech Inc C-Series Human Cytokine Antibody Array 9 Kit.
Supplier:  Raybiotech Inc AAHCYT92
Encompass_Preferred
Detects 51 Human Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: Adipsin (Complement Factor D), BCAM, CD30 (TNFRSF8), CD40 (TNFRSF5), Fc gamma RIIB/C (CD32B/C), Ferritin, FLRG, Follistatin, Furin, Galectin-7, GDF-15, Growth Hormone, IL-10 R alpha, IL-22, IL-28A (IFN-lambda 2), IL-29 (IFN-lambda 1), IL-31, Insulin, Luteinizing hormone, LIMPII, LYVE-1, Marapsin, MICA, MICB, MMP-2, MMP-7, MMP-8, MMP-10, NCAM-1 (CD56), Nidogen-1, NrCAM, NRG1-beta 1 (HRG1-beta 1), Osteopontin (SPP1), PAI-1, Platelet factor 4 (CXCL4), PSA-total, RAGE, RANK (TNFRSF11A), Resistin, SAA (Serum Amyloid A), Siglec-9, TACE, TIM-1 (KIM-1), TRAIL R2 (TNFRSF10B/DR5), Trappin-2, TREM-1, TSH, TSLP, VCAM-1 (CD106), VEGF-C, XEDAR
Catalog No. 50-194-8568