Learn More
Raybiotech Inc G-Series Human Cytokine Antibody Array 1000 Kit.
Catalog No. 501948572
Encompass_Preferred
A combination of Human Cytokine Antibody Array G6 & G7. Detects 120 Human Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. View protein list on RayBiotech’s website – AAH-CYT-G1000-8
Catalog No. 50-194-8572 Supplier Raybiotech Inc Supplier No. AAHCYTG10008
A combination of Human Cytokine Antibody Array G6 & G7. Detects 120 Human Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. View protein list on RayBiotech’s website – AAH-CYT-G1000-8