Learn More
Raybiotech Inc G-Series Human Cytokine Antibody Array 2000 Kit.
Catalog No. 501948573
Encompass_Preferred
A combination of Human Cytokine Antibody Array G6, G7, & G8. Detects 174 Human Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. View protein list on RayBiotech’s website – AAH-CYT-G2000-4
Catalog No. 50-194-8573 Supplier Raybiotech Inc Supplier No. AAHCYTG20004
A combination of Human Cytokine Antibody Array G6, G7, & G8. Detects 174 Human Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. View protein list on RayBiotech’s website – AAH-CYT-G2000-4