Learn More
Raybiotech Inc G-Series Human Cytokine Antibody Array 6 Kit.
Supplier:  Raybiotech Inc AAHCYTG68
Encompass_Preferred
Detects 60 Human Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. View protein list on RayBiotech’s website – AAH-CYT-G6-8
Catalog No. 50-194-8582