Learn More
Raybiotech Inc G-Series Human Cytokine Array 1 Kit.
Catalog No. 501950382
Encompass_Preferred
Semi-quantitative measurement of 20 human cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: GM-CSF, GRO alpha/beta/gamma, IFN-gamma, IL-1 alpha (IL-1 F1), IL-1 beta (IL-1 F2), IL-10, IL-12 p70, IL-13, IL-2, IL-4, IL-5, IL-6, IL-8 (CXCL8), MCP-1 (CCL2), MIP-1 alpha (CCL3), MIP-1 beta (CCL4), MMP-9, RANTES (CCL5), TNF alpha, VEGF-A
Catalog No. 50-195-0382 Supplier Raybiotech Inc Supplier No. GSHCYT11
Semi-quantitative measurement of 20 human cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: GM-CSF, GRO alpha/beta/gamma, IFN-gamma, IL-1 alpha (IL-1 F1), IL-1 beta (IL-1 F2), IL-10, IL-12 p70, IL-13, IL-2, IL-4, IL-5, IL-6, IL-8 (CXCL8), MCP-1 (CCL2), MIP-1 alpha (CCL3), MIP-1 beta (CCL4), MMP-9, RANTES (CCL5), TNF alpha, VEGF-A