Learn More
Raybiotech Inc G-Series Human Cytokine Array 2000 Kit.
Catalog No. 501950373
Encompass_Preferred
A combination of 3 non-overlapping arrays to measure the relative expression levels of 120 human cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. View protein list on RayBiotech’s website – GSH-CAA-2000-2
Catalog No. 50-195-0373 Supplier Raybiotech Inc Supplier No. GSHCAA20002
A combination of 3 non-overlapping arrays to measure the relative expression levels of 120 human cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. View protein list on RayBiotech’s website – GSH-CAA-2000-2