Learn More
Raybiotech Inc G-Series Human Cytokine Array 3000 Kit.
Supplier:  Raybiotech Inc GSHCAA30002
Encompass_Preferred
A combination of 4 non-overlapping arrays to measure the relative expression levels of 160 human cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. View protein list on RayBiotech’s website – GSH-CAA-3000-2
Catalog No. 50-195-0375