Learn More
Raybiotech Inc G-Series Human Cytokine Array 6 Kit.
Supplier:  Raybiotech Inc GSHCYT61
Encompass_Preferred
Semi-quantitative measurement of 40 human cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: 2B4 (CD244), ADAM-9, Angiopoietin-2, APRIL, BMP-2, BMP-9, C5a, Cathepsin L, CD200, CD97, Chemerin, DcR3, FABP2, FAP, FGF-19, Galectin-3, HGFR, IFN-alpha / beta R2, IGF-2, IGF-2 R, IL-1 R6 (IL-1 Rrp2), IL-24, IL-33 (IL-1 F11), Kallikrein 14, Legumain
Catalog No. 50-195-0388