Learn More
Raybiotech Inc G-Series Human Cytokine Array 9 Kit.
Catalog No. 501950394
Encompass_Preferred
Semi-quantitative measurement of 40 human cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: ADAMTS-13, Aggrecan, Angiotensinogen (SERPINA8), B7-H1 (CD274), BMPR-IA (ALK-3), BMPR-II, Cadherin-11, CD27 (TNFRSF7), CD6, Ck beta 8-1 (CCL23), CNTF, DNAM-1 (CD226), CD147 (EMMPRIN), FLRG, Follistatin-like 1, Fractalkine (CX3CL1), Galectin-1, GITR Ligand (TNFSF18), Granulysin (NKG5/LAG-2), IL-1 R3 (IL-1 R Acp), IL-15 R alpha, IL-17E (IL-25), IL-32 alpha, CHL-1 (L1CAM-2), LRIG3, LRP-6, MEPE (OF45), Nectin-4, Periostin, Persephin, Renin, RGM-B, ROBO3, S100 A8
Catalog No. 50-195-0394 Supplier Raybiotech Inc Supplier No. GSHCYT91
Semi-quantitative measurement of 40 human cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: ADAMTS-13, Aggrecan, Angiotensinogen (SERPINA8), B7-H1 (CD274), BMPR-IA (ALK-3), BMPR-II, Cadherin-11, CD27 (TNFRSF7), CD6, Ck beta 8-1 (CCL23), CNTF, DNAM-1 (CD226), CD147 (EMMPRIN), FLRG, Follistatin-like 1, Fractalkine (CX3CL1), Galectin-1, GITR Ligand (TNFSF18), Granulysin (NKG5/LAG-2), IL-1 R3 (IL-1 R Acp), IL-15 R alpha, IL-17E (IL-25), IL-32 alpha, CHL-1 (L1CAM-2), LRIG3, LRP-6, MEPE (OF45), Nectin-4, Periostin, Persephin, Renin, RGM-B, ROBO3, S100 A8