Learn More
Raybiotech Inc Quantibody Human Cytokine Array 2000 Kit.
Supplier:  Raybiotech Inc QAHCAA20001
Encompass_Preferred
A combination of Human Inflammation Array Q3, Human Growth Factor Array Q1, and Human Chemokine Array Q1. Detects 120 Human Inflammatory Factors, Growth Factors, and Chemokines. Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. View protein list on RayBiotech’s website – QAH-CAA-2000-1
Catalog No. 50-195-0805