Learn More
Raybiotech Inc Quantibody Human Cytokine Array 3000 Kit.
Supplier:  Raybiotech Inc QAHCAA30001
Encompass_Preferred
A combination of Human Inflammation Array Q3, Human Growth Factor Array Q1, Human Chemokine Array Q1, and Human Receptor Array Q1. Detects 160 Human Inflammatory Factors, Growth Factors, Chemokines and Receptors. Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. View protein list on RayBiotech’s website – QAH-CAA-3000-1
Catalog No. 50-195-0807