Learn More
Raybiotech Inc Quantibody Human Cytokine Array 4 Kit.
Supplier:  Raybiotech Inc QAHCYT41
Encompass_Preferred
Detects 40 Human Cytokines. Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. Proteins detected: Activin A, AgRP, Angiogenin, Angiopoietin-1, Angiostatin, Cathepsin S, CD40 (TNFRSF5), Cripto-1, DAN, DKK-1, E-Cadherin, Fas Ligand (TNFSF6), Fc gamma RIIB/C (CD32B/C), Follistatin, Galectin-7, gp130, ICAM-2 (CD102), IL-1 R4 (ST2), IL-13 R alpha 2, IL-13 R1, IL-17B, IL-2 R alpha, IL-2 R beta (CD122), IL-23, LAP/TGF beta 1, NrCAM, PAI-1, PDGF-AB, Resistin, SDF-1 beta (CXCL12 beta), Siglec-5 (CD170), Sonic Hedgehog N-Terminal (Shh-N), TGF beta 2, Thrombopoietin (TPO), Tie-2, TRAIL R4 (TNFRSF10D), TREM-1, TROP1 (EpCAM), VEGF-C, VEGFR1
Catalog No. 50-195-0817