Learn More
Raybiotech Inc Quantibody Human Cytokine Array 5 Kit.
Supplier:  Raybiotech Inc QAHCYT52
Encompass_Preferred
Detects 40 Human Cytokines. Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. Proteins detected: Adiponectin (ACRP30), Adipsin (Complement Factor D), Alpha-fetoprotein, ANGPTL4, Beta-2 Microglobulin, BCAM, CA125, CA15-3, CEA, CRP (C-Reactive Protein), ErbB2, Ferritin, FSH, GRO alpha (CXCL1), HCG beta, IGF-1 R, IL-1 R2, IL-3, IL-18 R beta (AcPL), IL-21, Leptin, MMP-1, MMP-2, MMP-3, MMP-8, MMP-9, MMP-10, MMP-13, NCAM-1 (CD56), Nidogen-1, NSE, Oncostatin M, Procalcitonin, Prolactin, PSA-free, Siglec-9, TACE, Thyroglobulin, TIMP-4, TSH
Catalog No. 50-195-0820