Learn More
Raybiotech Inc Quantibody Human Cytokine Array 6 Kit.
Supplier: Raybiotech Inc QAHCYT62
Detects 40 Human Cytokines. Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. Proteins detected: 2B4 (CD244), ADAM-9, Angiopoietin-2, APRIL, BMP-2, BMP-9, C5a, Cathepsin L, CD200, CD97, Chemerin, DcR3, FABP2, FAP, FGF-19, Galectin-3, HGFR, IFN-alpha / beta R2, IGF-2, IGF-2 R, IL-1 R6 (IL-1 Rrp2), IL-24, IL-33 (IL-1 F11), Kallikrein 14, Legumain