Learn More
Raybiotech Inc Quantibody Human Cytokine Array 7 Kit.
Supplier:  Raybiotech Inc QAHCYT72
Encompass_Preferred
Detects 40 Human Cytokines. Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. Proteins detected: ACE-2, aFGF (FGF-1), Albumin, AMICA, Angiopoietin-4, BAFF, CA19-9, CD163, Clusterin, CRTAM, CXCL14 (BRAK), Cystatin C, Decorin, Dkk-3, DLL1, Fetuin A, FOLR1, Furin, GASP-1 (WFIKKNRP), GASP-2 (WFIKKN), GCSF R (CD114), HAI-2, IL-17 RB, IL-27, LAG-3, LDL R, Pepsinogen 1, RANK (TNFRSF11A), RBP4, SOST, Syndecan-1, TACI (TNFRSF13B), TFPI, Thrombospondin 1, TRAIL R2 (TNFRSF10B/DR5), TRANCE (TNFSF11), Troponin I, uPA, VE-Cadherin (CDH5), WISP-1 (CCN4)
Catalog No. 50-195-0824