Learn More
Raybiotech Inc Quantibody Human Cytokine Array 8 Kit.
Supplier:  Raybiotech Inc QAHCYT81
Encompass_Preferred
Detects 40 Human Cytokines. Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. Proteins detected: ANGPTL3, beta IG-H3, CA9, Cathepsin B, CD23, CD26 (DPPIV), CHI3L1, Coagulation Factor III (Tissue Factor/Thromboplastin), CTLA-4 (CD152), Dkk-4, EDA-A2, Erythropoietin R, FGF-6, FGF-9, FOLH1 (PSMA), Gas 1, IGFBP-5, IL-1 F10 (IL-1HY2), IL-1 F5 (FIL1 delta), IL-1 F6 (FIL1 epsilon), IL-1 F7 (FIL1 zeta), IL-1 F8 (FIL1 eta), IL-1 F9 (IL-1 H1), IL-1 R5 (IL-18 R alpha), IL-10 R alpha, IL-17C, IL-18, IL-20, IL-34, IL-5 R alpha, Layilin, Leptin R, Marapsin, Mer, MMP-7, P-Cadherin, Prostasin, SIGIRR, TGF beta R3, TWEAK (TNFSF12)
Catalog No. 50-195-0825