Learn More
Creative Biomart Talimogene laherparepvec; T-Vec
Catalog No. 502084055
Encompass_Preferred
The product is a genetically administered herpes simplex virus 1 (HSV-1) that expresses human cytokine granulocyte-macrophage colony stimulating factor (GM-CSF) with antitumor and immune-stimulating activities. It specifically replicates within tumor cells and causes lysis.
Catalog No. 50-208-4055 Supplier Creative Biomart Supplier No. THP0004
The product is a genetically administered herpes simplex virus 1 (HSV-1) that expresses human cytokine granulocyte-macrophage colony stimulating factor (GM-CSF) with antitumor and immune-stimulating activities. It specifically replicates within tumor cells and causes lysis.