Learn More
Biomatik Corporation Human Cytoplasmic Antiproteinase 3 (CAP3) ELISA Kit
Supplier:  Biomatik Corporation EKU03620
Encompass_Preferred
Human Cytoplasmic Antiproteinase 3 (CAP3) ELISA Kit for detecting CAP3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.058ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P50453. Gene ID: 5272. Target Synonyms: SERPINB9; PI9; Serpin Peptidase Inhibitor Clade B Member 9; Peptidase inhibitor 9. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4930