Learn More
Bioworld HUMAN CYTOTOXIC T-LYMPHOCYT
Catalog No. 50253144
Encompass_Preferred
HUMAN CYTOTOXIC T-LYMPHOCYTE ASSOCIATED ANTIGEN-4 (RECOMBINANT): Recombinant Human Cytotoxic T-Lymphocyte Associated Antigen-4/Fc Chimera (CTLA-4) Source: Sf9 cells Background: CTLA-4 is expressed in low copy number by T-cells only a...
Catalog No. 50-253-144 Supplier Bioworld Supplier No. 220600461
HUMAN CYTOTOXIC T-LYMPHOCYTE ASSOCIATED ANTIGEN-4 (RECOMBINANT): Recombinant Human Cytotoxic T-Lymphocyte Associated Antigen-4/Fc Chimera (CTLA-4) Source: Sf9 cells Background: CTLA-4 is expressed in low copy number by T-cells only a...