Learn More
Biomatik Corporation Human Cytotoxic T-Lymphocyte Associated Antigen 4 (CTLA4) ELISA Kit
Supplier:  Biomatik Corporation EKU03621
Encompass_Preferred
Human Cytotoxic T-Lymphocyte Associated Antigen 4 (CTLA4) ELISA Kit for detecting CTLA4 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P16410. Gene ID: 1493. Target Synonyms: CD152; CELIAC3; CTLA4; GSE; IDDM12; Celiac Disease 3; Cytotoxic T-lymphocyte protein 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6523