Learn More
Abcam Human D-Dimer, Fluorescent ELISA Kit, 1 x 96 Tests
Catalog No. 502698357
Encompass_Preferred
Human D-Dimer, Fluorescent ELISA Kit is a single-wash 90-min Simplestep used to quantify Human D-Dimer with a sensitivity of 0.065 ng/mL. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Fluorescent Sandwich ELISA - 530/570/590 nm readout : works on any standard plate reader
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-269-8357 Supplier Abcam Supplier No. AB3153111X96TESTS
Add to Cart
Add to Cart
Human D-Dimer, Fluorescent ELISA Kit is a single-wash 90-min Simplestep used to quantify Human D-Dimer with a sensitivity of 0.065 ng/mL. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Fluorescent Sandwich ELISA - 530/570/590 nm readout : works on any standard plate reader
The product is subject to the following: Abcam Restricted Use Statement