Learn More
American Research Products Inc Human D Lactate Dehydrogenase
Catalog No. 5000329019
Encompass
Human D-Lactate Dehydrogenase D-LDH ELISA Kit from CUSABIO Detection Range 31 25 mIU/mL-2000 mIU/mL Sensitivity 7 8 mIU/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29019 Supplier American Research Products Inc Supplier No. CSBE11720H
Human D-Lactate Dehydrogenase D-LDH ELISA Kit from CUSABIO Detection Range 31 25 mIU/mL-2000 mIU/mL Sensitivity 7 8 mIU/mL Sample serum plasma tissue homogenates Method Sandwich