Learn More
Biomatik Corporation Human D2-Hydroxyglutarate Dehydrogenase (D2HGDH) ELISA Kit
Supplier:  Biomatik Corporation EKU03624
Encompass_Preferred
Human D2-Hydroxyglutarate Dehydrogenase (D2HGDH) ELISA Kit for detecting D2HGDH in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.065ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8N465. Gene ID: 728294. Target Synonyms: D2HGD. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6525