Learn More
Bioworld HUMAN D4-GDI (RECOMBINANT)
Supplier:  Bioworld 220603041
Encompass_Preferred
Recombinant Human rho-GDI (D4-GDI) Source: E.Coli Background: D4-GDI belongs to the family of rho-GDI or GDP-dissociation inhibitors for the Rho/Rac family of small G proteins; consisting of rhoGDI-1, D4-GDI/rhoGDI-2 and rhoGDI-3. It...
Catalog No. 50-253-403