Learn More
Biomatik Corporation Human D-amino Acid Oxidase Activator (DAOA) CLIA Kit
Supplier:  Biomatik Corporation EKU09489
Encompass_Preferred
Human D-amino Acid Oxidase Activator (DAOA) CLIA Kit for detecting DAOA in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 13.7-10,000pg/ml. Sensitivity: 6.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P59103. Gene ID: 267012. Target Synonyms: G72; LG72; SG72. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4577