Learn More
Biomatik Corporation Human D-Amino Acid Oxidase (DAO) CLIA Kit
Supplier:  Biomatik Corporation EKU09490
Encompass_Preferred
Human D-Amino Acid Oxidase (DAO) CLIA Kit for detecting DAO in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 13.7-10,000pg/ml. Sensitivity: 5.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P14920. Gene ID: 1610. Target Synonyms: DAMOX. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7724