Learn More
Abcam Human DAP12 (pY91) + DAP12 Total ELISA Kit, 1 x 96 Tests
Catalog No. 502703342
Encompass_Preferred
Human DAP12 (pY91) + DAP12 Total ELISA Kit is a single-wash 90-min Simplestep used to detect Human DAP12 (pY91) + DAP12 Total. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Semi-quantitative Sandwich ELISA - 450 nm readout : works on any standard plate reader
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-3342 Supplier Abcam Supplier No. AB3161361X96TESTS
Human DAP12 (pY91) + DAP12 Total ELISA Kit is a single-wash 90-min Simplestep used to detect Human DAP12 (pY91) + DAP12 Total. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Semi-quantitative Sandwich ELISA - 450 nm readout : works on any standard plate reader
The product is subject to the following: Abcam Restricted Use Statement