Learn More
Sino Biological Human DC-SIGN/CD209 Matched ELISA Antibody Pair Set 5 Plates
Catalog No. 5000785893
Encompass_Preferred
The Human DC-SIGN / CD209 ELISA Pair Set is for the quantitative determination of Human DC-SIGN / CD209. This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for DC-SIGN / CD209 coated on a 96-well plate. Standards and samples are added to the wells, and any DC-SIGN / CD209 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-DC-SIGN / CD209 monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of DC-SIGN / CD209 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-85893 Supplier Sino Biological Supplier No. SEK102005
The Human DC-SIGN / CD209 ELISA Pair Set is for the quantitative determination of Human DC-SIGN / CD209. This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for DC-SIGN / CD209 coated on a 96-well plate. Standards and samples are added to the wells, and any DC-SIGN / CD209 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-DC-SIGN / CD209 monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of DC-SIGN / CD209 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.