Learn More
BIORTUS USA INC Human DCAF16/DDB1 Protein, 100UG
Catalog No. NC2953301
Encompass_Preferred
Gene Name: DCAF16/DDB1. Uniprot: Q9NV06/Q16531. Construct:, His-TEV-G-DCAF16(G2-L216 end)/DDB1(M1-H1140 end). Expression Host: Insect cell. Purity: 90%.
Catalog No. NC2953301 Supplier BIORTUS USA INC Supplier No. BP7872D01A
Add to Cart
Add to Cart
Gene Name: DCAF16/DDB1. Uniprot: Q9NV06/Q16531. Construct:, His-TEV-G-DCAF16(G2-L216 end)/DDB1(M1-H1140 end). Expression Host: Insect cell. Purity: 90%.