Learn More
BIORTUS USA INC Human DCAF2/DDB1 Protein, 100UG
Catalog No. NC2953308
Encompass_Preferred
Gene Name: DTL/DDB1. Uniprot: Q9NZJ0/Q16531. Construct:, His-Strep II-TEV-(GG)-DCAF2(L2-L730 end)/DDB1(M1-H1140 end). Expression Host: Insect cell. Purity: 90%.
Catalog No. NC2953308 Supplier BIORTUS USA INC Supplier No. BP2706313D
Gene Name: DTL/DDB1. Uniprot: Q9NZJ0/Q16531. Construct:, His-Strep II-TEV-(GG)-DCAF2(L2-L730 end)/DDB1(M1-H1140 end). Expression Host: Insect cell. Purity: 90%.