Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human DCBLD2/ESDN ELISA Kit

Catalog No. EH148RBX10
Encompass_Preferred
Change view
Click to view available options
10 x 96 Tests; HRP
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
EH148RBX10 10 x 96 Tests; HRP
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EH148RBX10 Supplier Invitrogen™ Supplier No. EH148RBX10
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

ELISA

Human DCBLD2/ESDN quantitates human DCBLD2/ESDN in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human DCBLD2/ESDN.

This protein is a key enzyme in the catabolic pathway of odd-chain fatty acids, isoleucine, threonine, methionine, and valine. It is probably a dodecamer composed of six biotin-containing alpha subunits and six beta subunits. The protein is located in the mitochondrial matrix. Defects in PCCA are the cause o fpropionic acidemia type I (PA-1) [MIM:606054]. PA-1 is a life-threatening disease characterized by episodic vomiting, lethargy and ketosis, neutropenia, periodic thrombocytopenia, hypogammaglobulinemia, develop-mental retardation, and intolerance to protein.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q96PD2
Assay Range 0.041-10 ng/mL
Assay Sensitivity 0.041 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias 1700055P21Rik; AW146002; CLCP1; coagulation factor V/VIII-homology domains protein 1; CUB, LCCL and coagulation factor V/VIII-homology domains protein 1; DCBLD2; discoidin; discoidin, CUB and LCCL domain containing 2; discoidin, CUB and LCCL domain-containing protein 2; Endothelial and smooth muscle cell-derived neuropilin-like protein; ESDN1700055P21Rik; AW146002; CLCP1; coagulation factor V/VIII-homology domains protein 1; CUB, LCCL and coagulation factor V/VIII-homology domains protein 1; DCBLD2; discoidin; discoidin, CUB and LCCL domain containing 2; discoidin, CUB and LCCL domain-containing protein 2; Endothelial and smooth muscle cell-derived neuropilin-like protein; ESDN;
Gene ID (Entrez) 131566
Gene Symbol DCBLD2
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target DCBLD2
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.