Learn More
Raybiotech Inc Human DCBLD2/ESDN ELISA Kit
Supplier:  Raybiotech Inc ELHDCBLD21
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 0.041 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects Human DCBLD2
Catalog No. 50-194-9180