Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human DCLK3 Synthetic Peptide

Catalog No. PEP1025
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP1025 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP1025 Supplier Invitrogen™ Supplier No. PEP1025
Add to Cart
Add to Cart

Synthetic Peptide

This peptide corresponds to 14 amino acids near the carboxy terminus of human DCLK3. PEP-1025 can be used as a blocking peptide with polyclonal antibody PA5-20911.

DCLK3 is one of three doublecortin-like kinases similar to the Ca2+/calmodulin-dependent protein kinase (CaMK) family. DCLK3 mRNA, like that of the homologous DCLK1 and DCLK3, is highly expressed in adult brain, but only DCLK3 transcripts are present in liver and kidney, suggesting that DCLK3 may play other roles than in cortical development. The DCLK proteins are homologous to Doublecortin (DCX), a protein that is mutated in X-linked human lissencephaly. In mouse models where the DCX gene has been disrupted, DCLK1 expression increases slightly and appears to compensate for the loss of DCX, as mice mutant for both DCX and DCLK1 show a severe phenotype including perinatal lethality, disorganized neocortical layering, and profound hippocampal cytoarchitectural disorganization.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 85443
Species Human
Purification Method purified
Accession Number Q9C098
Common Name DCLK3
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Alias BC056929; C730036H08; CLICK-I and II-related; Click-I, -II related; CLICK-I,II-related; CLR; Dcamkl3; DCDC3C; DCK3; Dclk3; doublecortin and CaM kinase-like 3; doublecortin domain-containing protein 3C; doublecortin like kinase 3; doublecortin-like and CAM kinase-like 3; Doublecortin-like kinase 3; hCG_16695; KIAA1765; RGD1309232; serine/threonine-protein kinase DCLK3
Gene Symbol DCLK3
Lyophilized No
Preservative Sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.