Learn More
Raybiotech Inc Human DcR3 ELISA Kit
Supplier:  Raybiotech Inc ELHDCR31
Encompass_Preferred
For cell culture supernatants, EDTA and/or Citrate treated plasma, and serum samples. Heparin is not recommended as an anticoagulant For use in this assay. Sensitivity: 0.3 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects human DcR3. Other species not determined.
Catalog No. 50-194-9181