Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems Human DcR3 (TNFRSF6B) Recombinant Protein, His Tag, HEK293-expressed - 50 μg

Catalog No. NC3718221
Encompass_Preferred

Recombinant human DcR3 (TNFRSF6B) extracellular domain expressed in HEK293 cells with a C-terminal His tag. Contains amino acids Val 30-Ser 195 (UniProt O95407). Supplied as 50 μg.

  • Expression system: HEK293 (human 293) cells.
  • Amino acid range: Val 30-Ser 195.
  • UniProt accession: O95407.
  • Tag: C-terminal His tag.
  • Pack size: 50 μg.

Catalog No. NC3718221 Supplier ACROBiosystems Supplier No. DC3H52H350UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human DcR3 (TNFRSF6B) extracellular domain expressed in HEK293 cells with a C-terminal His tag. Contains amino acids Val 30-Ser 195 (UniProt O95407). Supplied as 50 μg.

  • Expression system: HEK293 (human 293) cells.
  • Amino acid range: Val 30-Ser 195.
  • UniProt accession: O95407.
  • Tag: C-terminal His tag.
  • Pack size: 50 μg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.