Learn More
Sino Biological Anti-DCTN4 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 200665T44100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human DCTN4 fragment, and purified by antigen affinity chromatography.
Catalog No. 50-240-9027