Learn More
BioVendor, LLC Human DD/E FDP Plm Generated, Mouse Monoclonal Antibody (500 ug)
SDP
Supplier:  BioVendor, LLC QZBFBDDE2500
Encompass
Human DD/E FDP Plm Generated, Mouse Monoclonal Antibody (500 ug). Manufactured by QuickZyme Biosciences. Mouse MAb against human DD/E complex (specific for plasmin generated fibrin degredation products). Does not react with fibrinogen, fibrin, or non-cross linked fibrin(ogen) degradation products. Can be used in ELISA or agglutination test for such cross-linked degradation products. Synonyms: D-Dimer DD/E2 Fibrin Split Products, FSP. SUMMARY: Fibrin functions in hemostasis as a primary component of blood clots. It is also important in the early stages of wound repair. Fibrinolysis is the enzymatic breakdown of fibrin in blood clots and results in various Fibrin Degradation Products (FDP). High levels of FDP in plasma indicate abnormal fibrinolytic and/or fibrinogenolytic activity. Elevated FDP levels are typical in disseminated intravascular coagulation (DIC), but are not specific for DIC and are also seen numerous other clinical conditions.
Catalog No. 50-206-9099