Learn More
Sino Biological Human DDC MAb 100ul (Sino Biological)
Supplier:  Sino Biological 10560R005100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human DOPA Decarboxylase / DDC (rh DOPA Decarboxylase / DDC; Catalog#10560-H08B; NP_000781.1; Met 1-Glu 480).
Catalog No. 50-237-3697