Learn More
Sino Biological Anti-DOPA Decarboxylase/DDC Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10560RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant full length of human DDC / AADC (rh DDC; Catalog#10560-H08B; NP_000781.1; Met 1-Glu 480). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-0695