Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DDEFL1 Full-length ORF (NP_060177.2, 1 a.a. - 903 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. p-7101105 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-941-582 25 μg
89-941-581 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-941-582 Supplier Abnova™ Supplier No. H00055616P01L
Only null left
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a member of a subfamily of ADP-ribosylation factor(Arf) GTPase-activating proteins that contain additional ankyrin repeat and pleckstrin homology domains. The Arf GAP domain of this protein catalyzes the hydrolysis of GTP bound to Arf proteins. The encoded protein promotes cell differentiation and migration and has been implicated in cancer cell invasion. Alternative splicing results in multiple transcript variants. [provided by RefSeq]

Sequence: MPEQFSVAEFLAVTAEDLSSPAGAAAFAAKMPRYRGAALAREEILEGDQAILQRIKKAVRAIHSSGLGHVENEEQYREAVESLGNSHLSQNSHELSTGFLNLAVFTREVAALFKNLIQNLNNIVSFPLDSLMKGQLRDGRQDSKKQLEKAWKDYEAKMAKLEKERDRARVTGGIPGEVAQDMQRERRIFQLHMCEYLLKAGESQMKQGPDFLQSLIKFFHAQHNFFQDGWKAAQSLFPFIEKLAASVHALHQAQEDELQKLTQLRDSLRGTLQLESREEHLSRKNSGCGYSIHQHQGNKQFGTEKVGFLYKKSDGIRRVWQKRKCGVKYGCLTISHSTINRPPVKLTLLTCQVRPNPEEKKCFDLVTHNRTYHFQAEDEHECEAWVSVLQNSKDEALSSAFLGEPSAGPGSWGSAGHDGEPHDLTKLLIAEVKSRPGNSQCCDCGAADPTWLSTNLGVLTCIQCSGVHRELGVRFSRMQSLTLDLLGPSELLLALNMGNTSFNEVMEAQLPSHGGPKPSAESDMGTRRDYIMAKYVEHRFARRCTPEPQRLWTAICNRDLLSVLEAFANGQDFGQPLPGPDAQAPEELVLHLAVKVANQASLPLVDFIIQNGGHLDAKAADGNTALHYAALYNQPDCLKLLLKGRALVGTVNEAGETALDIARKKHHKECEELLEQAQAGTFAFPLHVDYSWVISTEPGSDSEEDEEEKRCLLKLPAQAHWASGRLDISNKTYETVASLGAATPQGESEDCPPPLPVKNSSRTLVQGCARHASGDRSEVSSLSSEAPETPESLGSPASSSSLMSPLEPGDPSQAPPNSEEGLREPPGTSRPSLTSGTTPSEMYLPVRFSSESTRSYRRGARSPEDGPSARQPLPRRNVPVGITEGDGSRTGSLPASSVQLLQD

Specifications

Accession Number NP_060177.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 55616
Molecular Weight (g/mol) 125.6kDa
Name DDEFL1 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias ACAP4/CENTB6/DDEFL1/FLJ20199/UPLC1
Common Name ASAP3
Gene Symbol ASAP3
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.