Learn More
Biomatik Corporation Human DDT (D-Dopachrome Tautomerase) ELISA Kit
Supplier:  Biomatik Corporation EKF58778
Encompass_Preferred
Human DDT (D-Dopachrome Tautomerase) ELISA Kit for detecting DDT in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P30046. Target Synonyms: DDT; DDCT; D-Dopachrome Decarboxylase; DOPD; MIF2; Phenylpyruvate Tautomerase II; D-dopachrome tautomerase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5443