Learn More
Sino Biological Anti-DDX41 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 102459T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human DDX41, and purified by antigen affinity chromatography.
Catalog No. 50-241-1071