Learn More
Biomatik Corporation Human Death Associated Protein 3 (DAP3) ELISA Kit
Supplier:  Biomatik Corporation EKU10786
Encompass_Preferred
Human Death Associated Protein 3 (DAP3) ELISA Kit for detecting DAP3 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.114ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P51398. Gene ID: 7818. Target Synonyms: MRP-S29; MRPS29; bMRP-10; 28S Ribosomal Protein S29,Mitochondrial; Ionizing radiation resistance conferring protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8289