Learn More
Biomatik Corporation Human Death Inducer Obliterator 1 (DIDO1) ELISA Kit
Supplier:  Biomatik Corporation EKU03641
Encompass_Preferred
Human Death Inducer Obliterator 1 (DIDO1) ELISA Kit for detecting DIDO1 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.104ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9BTC0. Gene ID: 11083. Target Synonyms: BYE1; DATF1; DIDO2; DIDO3; DIO1; Death Associated Transcription Factor 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6844