Learn More
Raybiotech Inc Human Dectin-1 (CLEC7A) ELISA Kit
Supplier:  Raybiotech Inc ELHDECTIN11
Encompass_Preferred
For Cell Culture Supernatants, Plasma, and Serum samples. Sensitivity: 0.082 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human Dectin-1. Other species not yet determined.
Catalog No. 50-194-9186