Learn More
Biomatik Corporation Human Dedicator Of Cytokinesis 4 (DOCK4) ELISA Kit
Supplier:  Biomatik Corporation EKU03646
Encompass_Preferred
Human Dedicator Of Cytokinesis 4 (DOCK4) ELISA Kit for detecting DOCK4 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.063ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8N1I0. Gene ID: 9732. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9017