Learn More
Sino Biological Anti-beta Defensin 3/DEFB103A Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10849T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Defensin / Beta-defensin 3 / DEFB103 (rh Defensin / Beta-defensin 3 / DEFB103 ; Catalog#10849-H07E; P81534; Gly23-Lys6). Defensin / Beta-defensin 3 / DEFB103 specific IgG was purified by Human Defensin / Beta-defensin 3 / DEFB103 affinity chromatography.
Catalog No. 50-255-5129