Learn More
Sino Biological Recombinant Human beta Defensin 3/DEFB103A Protein (His Tag)
Supplier:  Sino Biological 10849H07E1
Encompass_Preferred
A DNA sequence encoding the mature form of human DEFB103A (P81534) (Gly 23-Lys 67) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-255-5130